Skip to Content
Merck

Skip To

810682

Probumin® Bovine Serum Albumin Media Grade

pkg of 10 g, ≥98%, USA origin, Tested for IgG, microbiological culture, microbiological culture: suitable, Binding, transport studies

Synonym(s):

Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

Estimated to ship on26 June 2028fromAreál Kühne+Nagel spol. s r.o.

385,00 CZK

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801
NACRES:
NA.28
Assay:
≥98% (purity)
Form:
lyophilized powder
Technique(s):
microbiological culture: suitable
Impurities:
Tested for IgG

385,00 CZK


Estimated to ship on26 June 2028Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Probumin® Bovine Serum Albumin Media Grade, media grade, pkg of 10 g

Quality Segment

assay

≥98% (purity)

form

lyophilized powder

shelf life

48 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pkg of 10 g

manufacturer/tradename

Probumin®

origin

USA origin

technique(s)

microbiological culture: suitable

dilution

(Binding, transport studies)

impurities

Tested for IgG

loss

≤5%

pH

6.8-7.2

anion traces

chloride (Cl-): 0-3.0 mg/g

cation traces

Na: 0-10 mg/g

foreign activity

Tested for Endotoxin , Tested for mycoplasma

General description

Albumin is one of the most extensively utilized proteins in the medical industry. Its availability, inherent stability, and ligand binding capability make albumin a critical raw material in the development and production of many diagnostic tests. With over 65 years of protein fractionation expertise, we are an experienced manufacturer of consistent bovine serum albumin products for the diagnostic and cell culture markets. Our proprietary heat shock and cold solvent fractionation processing technology allows for the delivery of products with high-quality, performance, and reliability.

Application

Probumin® BSA products are fit for use as blockers, protein/antibody (conjugate) stabilizers, and in immunoassays (EIA, RIA and serology systems).

Features and Benefits

  • High-quality and reliability. Assurance of consistency in performance and supply.
  • Proprietary heat shock and cold solvent fractionation process optimizes the protein yield, purity, and efficacy of BSA. Large batch capacity.
  • Safe serum sourcing from USDA-licensed facilities exclusively from U.S. cattle.

Preparation Note

Storage temperature< -10⁰ C

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
820453820456820464
description

media grade, pkg of 10 g

description

lyophilized powder, pkg of 10 g

description

lyophilized powder, pkg of 5 kg

description

solution (30%), pkg of 50 mL

technique(s)

microbiological culture: suitable

technique(s)

ELISA: suitable, enzyme immunoassay: suitable

technique(s)

ELISA: suitable, enzyme immunoassay: suitable

technique(s)

ELISA: suitable, enzyme immunoassay: suitable

origin

USA origin

origin

USA origin

origin

USA origin

origin

USA origin

assay

≥98% (purity)

assay

≥98% (purity)

assay

≥98% (purity)

assay

29-31 g/dL protein basis (biuret)

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

solution (30%)

impurities

Tested for IgG

impurities

Tested for IgG

impurities

Tested for IgG

impurities

Tested for IgG


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters