Skip to Content
Merck

Skip To

8.08803

Zinc(II) acetylacetonate

for synthesis

Synonym(s):

Zinc(II) acetylacetonate, Bis(acetylacetonato)zinc(II), Bis(2,4-pentanedionato) zinc(II)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 2,120.00

About This Item

Empirical Formula (Hill Notation):
C10H14O4Zn
CAS Number:
Molecular Weight:
263.61
MDL number:
UNSPSC Code:
12352103
EC Index Number:
237-860-3
NACRES:
NA.22
Form:
powder

CZK 2,120.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1 hPa ( 20 °C)

Quality Segment

form

powder

autoignition temp.

580 °C

pH

10 (20 °C, 4 g/L in H2O)

bp

129-131 °C/13 hPa

mp

124-126 °C

solubility

6.9 g/L

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[Zn+2].O=C([C-H]C(=O)C)C.O=C([C-H]C(=O)C)C

InChI

1S/2C5H7O2.Zn/c2*1-4(6)3-5(2)7;/h2*3H,1-2H3;/q2*-1;+2

InChI key

NHXVNEDMKGDNPR-UHFFFAOYSA-N

Analysis Note

Assay (ex Zn): ≥ 95.0 %
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.089158.027128.08305
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

solubility

6.9 g/L

solubility

14 g/L

solubility

-

solubility

-

bp

129-131 °C/13 hPa

bp

-

bp

160 °C/13 hPa

bp

-

form

powder

form

powder

form

powder

form

powder

mp

124-126 °C

mp

189-192 °C

mp

245 °C (decomposition)

mp

200 °C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Hello, What is the purity of your product? Thanks, J.

    1 answer
    1. The purity of this product is specified as ≥ 95.0 %. The exact purity result of this product will be lot specific and reported on the certificate of analysis.

      Helpful?

Reviews

No rating value

Active Filters