Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 L | Please contact Customer Service for Availability | CZK 1,260.00 |
About This Item
vapor pressure
0.01 hPa ( 20 °C)
Quality Segment
assay
≥98.0% (GC)
form
liquid
autoignition temp.
340 °C (DIN 51794)
potency
>18056 mg/kg LD50, oral (Rat), >18056 mg/kg LD50, skin (Rabbit)
expl. lim.
0.5-3.4 % (v/v)
pH
8.5-9.0 (20 °C, 500 g/L in H2O)
kinematic viscosity
50.13 cSt(20.65 °C)
bp
327-328 °C/1013 hPa
mp
-9.4 °C
transition temp
flash point 204 °C
density
1.12 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O(CCOCCO)CCOCCO
InChI
1S/C8H18O5/c9-1-3-11-5-7-13-8-6-12-4-2-10/h9-10H,1-8H2
InChI key
UWHCKJMYHZGTIT-UHFFFAOYSA-N
Analysis Note
Appearance of substance (visual): clear
Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.121 - 1.124
Identity (IR): passes test
Water (K. F.): ≤ 0.3 %
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form liquid |
| assay ≥98.0% (GC) | assay ≥98.0% (GC) | assay ≥98.0% (GC) | assay ≥98% (GC) |
| Quality Level 200 | Quality Level 200 | Quality Level - | Quality Level 200 |
| storage temp. 2-30°C | storage temp. 2-30°C | storage temp. - | storage temp. - |
| mp -9.4 °C | mp -30 °C | mp - | mp - |
| density 1.12 g/cm3 at 20 °C | density 1.01 g/cm3 at 20 °C | density 0.977 g/mL at 20 °C | density 0.969 g/mL at 20 °C (lit.) |
Still not finding the right product?
Explore all of our products under Tetraethylene glycol
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
359.6 °F - Pensky-Martens closed cup
flash_point_c
182 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



