Skip to Content
Merck

8.08608

1,3,5-Tris(2-hydroxyethyl)cyanuric acid

for synthesis

Synonym(s):

1,3,5-Tris(2-hydroxyethyl)cyanuric acid, 1,3,5-Tris(2-hydroxyethyl)hexahydro-s-triazine-2,4,6-trione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H15N3O6
CAS Number:
Molecular Weight:
261.23
EC Index Number:
212-660-9
UNSPSC Code:
12352115
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98.0% (HPLC)

autoignition temp.

430 °C

potency

>10000 mg/kg LD50, oral (Rat)

pH

7 (20 °C, 100 g/L in H2O)

mp

134-138 °C

transition temp

flash point 270 °C

solubility

1150 g/L

bulk density

580 kg/m3

storage temp.

2-30°C

InChI

1S/C9H15N3O6/c13-4-1-10-7(16)11(2-5-14)9(18)12(3-6-15)8(10)17/h13-15H,1-6H2

InChI key

BPXVHIRIPLPOPT-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 134 °C
Melting range (upper value): ≤ 138 °C
Identity (IR): passes test




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library