Skip to Content
Merck

Skip To

8.08333

Methyl-4-toluenesulfonate

for synthesis

Synonym(s):

Methyl-4-toluenesulfonate, p-Toluenesulfonic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 712.00
500 g
Please contact Customer Service for Availability
CZK 2,640.00
2.5 kg
Please contact Customer Service for Availability
CZK 9,690.00
25 kg
Please contact Customer Service for Availability
CZK 26,900.00

About This Item

Linear Formula:
4-(CH3)C6H4SO3CH3
CAS Number:
Molecular Weight:
186.23
UNSPSC Code:
12352108
EC Index Number:
201-283-5
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
solid

CZK 712.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.13 kPa ( 20 °C)

Quality Segment

assay

≥98.0% (GC)

form

solid

potency

341 mg/kg LD50, oral (Rat)

bp

292 °C/1013 hPa

transition temp

flash point 130 °C

density

1.23 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(OC)c1ccc(cc1)C

InChI

1S/C8H10O3S/c1-7-3-5-8(6-4-7)12(9,10)11-2/h3-6H,1-2H3

InChI key

VUQUOGPMUUJORT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.08332132004158992
assay

≥98.0% (GC)

assay

≥97.0% (GC)

assay

97%

assay

98%

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

200

density

1.23 g/cm3 at 25 °C

density

1.174 g/cm3 at 20 °C

density

-

density

1.234 g/mL at 25 °C (lit.)

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

form

solid

form

solid

form

powder

form

solid

bp

292 °C/1013 hPa

bp

221 °C/1013 hPa

bp

-

bp

144-145 °C/5 mmHg (lit.)


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

wgk

WGK 2

flash_point_f

266.0 °F - closed cup

flash_point_c

130 °C - closed cup

Storage Class

8A - Combustible corrosive hazardous materials


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

108-88-3

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters