Skip to Content
Merck
All Photos(1)

Key Documents

8.07383

Sigma-Aldrich

Lead(II) chloride

anhydrous for synthesis

Synonym(s):

Lead(II) chloride, Lead dichloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 837.00
500 G
CZK 2,100.00

CZK 837.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
CZK 837.00
500 G
CZK 2,100.00

About This Item

Empirical Formula (Hill Notation):
Cl2Pb
CAS Number:
Molecular Weight:
278.11
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-845-5
NACRES:
NA.22
grade:
for synthesis
form:
powder

CZK 837.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for synthesis

Quality Level

form

powder

potency

>1947 mg/kg LD50, oral (Rat)

bp

950 °C/1013 hPa

mp

500 °C

density

5.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Pb+2].[Cl-].[Cl-]

InChI

1S/2ClH.Pb/h2*1H;/q;;+2/p-2

InChI key

HWSZZLVAJGOAAY-UHFFFAOYSA-L

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (ICP-OES): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1A - STOT RE 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service