Skip to Content
Merck

Skip To

8.03977

9-Fluorenone

for synthesis

Synonym(s):

9-Fluorenone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 g
Please contact Customer Service for Availability
CZK 2,510.00

About This Item

Empirical Formula (Hill Notation):
C13H8O
CAS Number:
Molecular Weight:
180.20
UNSPSC Code:
12352115
EC Index Number:
207-630-7
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Form:
solid

CZK 2,510.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (GC)

form

solid

autoignition temp.

608 °C

potency

>3900 mg/kg LD50, oral (Rat)

bp

342 °C/1013 hPa

mp

81-84 °C

transition temp

flash point 163 °C

density

0.9 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=[c]1c2c(c3c1cccc3)cccc2

InChI

1S/C13H8O/c14-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)13/h1-8H

InChI key

YLQWCDOCJODRMT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 81 °C
Melting range (upper value): ≤ 84 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.41569H31204F1557
assay

≥99.0% (GC)

assay

-

assay

96%

assay

97%

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

mp

81-84 °C

mp

>300 °C

mp

153-154 °C (lit.)

mp

225-227 °C (lit.)

density

0.9 g/cm3 at 20 °C

density

-

density

-

density

-

bp

342 °C/1013 hPa

bp

-

bp

-

bp

-

form

solid

form

powder

form

-

form

-


Still not finding the right product?

Explore all of our products under 9-Fluorenone


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

325.4 °F

flash_point_c

163 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters