Skip to Content
Merck
All Photos(2)

Key Documents

8.03456

Sigma-Aldrich

1,4-Diazabicyclo[2.2.2]octane

greener alternative

for synthesis

Synonym(s):

1,4-Diazabicyclo[2.2.2]octane, Triethylenediamine, DABCO

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 1,270.00
250 G
CZK 2,930.00
1 KG
CZK 9,270.00

CZK 1,270.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
CZK 1,270.00
250 G
CZK 2,930.00
1 KG
CZK 9,270.00

About This Item

Empirical Formula (Hill Notation):
C6H12N2
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352005
EC Index Number:
205-999-9
NACRES:
NA.22

CZK 1,270.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for synthesis

Quality Level

vapor pressure

0.68 hPa ( 21 °C)

Assay

≥98.0% (GC)

form

solid

autoignition temp.

350 °C

potency

700 mg/kg LD50, oral (Rat)

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

173.4 °C/1000 hPa

mp

155-158 °C

transition temp

flash point 62.2 °C

solubility

400 g/L

density

1.14 g/cm3 at 25 °C

bulk density

800 kg/m3

greener alternative category

storage temp.

2-30°C

SMILES string

N21CCN(CC2)CC1

InChI

1S/C6H12N2/c1-2-8-5-3-7(1)4-6-8/h1-6H2

InChI key

IMNIMPAHZVJRPE-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

  • Biocompatible Polymer Composites: Research demonstrates the development of a reactive compatibilization method using 1,4-diazabicyclo[2.2.2]octane as a catalyst for the preparation of polylactic acid/poly(butylene adipate-co-terephthalate)/thermoplastic starch ternary bio-composites. This approach significantly improves the mechanical and biodegradable properties of the composites, suitable for environmentally friendly applications in the packaging and automotive industries (Fang et al., 2024).
  • Cobalt Single-Atom Catalysts: Research on the development of cobalt single-atom catalysts encapsulated in a metal-organic framework using 1,4-diazabicyclo[2.2.2]octane. This catalyst shows exceptional efficiency in the oxygen reduction reaction, important for energy conversion technologies such as fuel cells and metal-air batteries (Gao et al., 2024).
  • Advanced Analytical Techniques: Study on the use of 1,4-diazabicyclo[2.2.2]octane in advanced derivatization techniques for the analysis of Novichok agents in biofluids using LC-MS. This research contributes to the field of forensic science and chemical warfare agent detection, providing robust methods for emergency responses and public safety (Yamaguchi et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Water (K. F.): ≤ 1.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

144.0 °F - closed cup

Flash Point(C)

62.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service