Skip to Content
Merck

Skip To

8.03259

Diphenylacetonitrile

for synthesis

Synonym(s):

Diphenylacetonitrile, Benzhydryl cyanide, Diphenylmethyl cyanide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 294.00

About This Item

Linear Formula:
(C6H5)2CHCN
CAS Number:
Molecular Weight:
193.24
UNSPSC Code:
12352117
EC Index Number:
201-662-5
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (GC)
Form:
powder

CZK 294.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

21.3 hPa ( 190 °C)

Quality Segment

assay

≥99.0% (GC)

form

powder

potency

3500 mg/kg LD50, oral (Rat)

bp

175-177 °C/15 hPa

mp

72-75 °C

transition temp

flash point 120 °C

storage temp.

2-30°C

SMILES string

N#CC(c2ccccc2)c1ccccc1

InChI

1S/C14H11N/c15-11-14(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,14H

InChI key

NEBPTMCRLHKPOB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 72 °C
Melting range (upper value): ≤ 75 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.06437B4856248835
assay

≥99.0% (GC)

assay

≥95.0% (silver nitrate titration)

assay

99%

assay

95%

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

form

powder

form

solid

form

powder

form

solid

bp

175-177 °C/15 hPa

bp

1496 °C/1013 hPa

bp

297-298 °C (lit.)

bp

105-106 °C/1 mmHg (lit.)

mp

72-75 °C

mp

563 °C

mp

65-67 °C (lit.)

mp

47-50 °C (lit.)

storage temp.

2-30°C

storage temp.

no temp limit

storage temp.

-

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Diphenylacetonitrile


pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

245.5 °F - Pensky-Martens closed cup

flash_point_c

118.6 °C - Pensky-Martens closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters