Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
5 g | Please contact Customer Service for Availability | CZK 779.00 CZK 584.25 |
250 g | Please contact Customer Service for Availability | CZK 3,070.00 |
About This Item
CZK 584.25
List PriceCZK 779.00Save 25%vapor pressure
<0.001 hPa ( 25 °C)
Quality Segment
assay
≥99.0% (GC)
form
powder
autoignition temp.
>365 °C
pH
7 (1 g/L in H2O)
bp
>300 °C (decomposes)
mp
176-178 °C (lit.), 177-180 °C
transition temp
flash point 224 °C
solubility
1 g/L
bulk density
800 kg/m3
storage temp.
2-30°C
SMILES string
Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O
InChI
1S/C6H5N3O4/c7-5-2-1-4(8(10)11)3-6(5)9(12)13/h1-3H,7H2
InChI key
LXQOQPGNCGEELI-UHFFFAOYSA-N
Analysis Note
Melting range (lower value): ≥ 177 °C
Melting range (upper value): ≤ 180 °C
Identity (IR): passes test
1 of 1
This Item | ||
|---|---|---|
| assay ≥99.0% (GC) | assay - | assay 98% |
| solubility 1 g/L | solubility - | solubility - |
| Quality Level 200 | Quality Level - | Quality Level 100 |
| form powder | form solid | form - |
| storage temp. 2-30°C | storage temp. - | storage temp. - |
| mp 176-178 °C (lit.), 177-180 °C | mp - | mp 176-178 °C (lit.) |
Still not finding the right product?
Explore all of our products under 2,4-Dinitroaniline
signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2
Storage Class
6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
435.2 °F
flash_point_c
224 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




