Skip to Content
Merck

Skip To

8.01059

Aluminium acetylacetonate

for synthesis

Synonym(s):

Aluminium acetylacetonate, Tris(acetylacetonato)aluminium, Tris(2,4-pentanedionato)aluminium, Aluminium 2,4-pentanedionate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 918.00
250 g
Please contact Customer Service for Availability
CZK 2,930.00
25 kg
Please contact Customer Service for Availability
CZK 150,500.00
50 kg
Please contact Customer Service for Availability
CZK 293,600.00

About This Item

Empirical Formula (Hill Notation):
C15H21AlO6
CAS Number:
Molecular Weight:
324.31
UNSPSC Code:
12352103
EC Index Number:
237-741-6
NACRES:
NA.22
MDL number:

CZK 918.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

autoignition temp.

500 °C

potency

48.7 mg/kg LD50, oral (Rat)

pH

8 (20 °C, 3 g/L in H2O)

bp

315 °C/1013 hPa

mp

190-193 °C

solubility

3 g/L

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

[Al+3].[O-]\C(=C/C(=O)C)\C.[O-]\C(=C/C(=O)C)\C.[O-]\C(=C/C(=O)C)\C

InChI key

KILURZWTCGSYRE-LNTINUHCSA-K

Application

  • Viscoelasticity in Pressure-Sensitive Adhesives: This study examines the influence of crosslink density and linear viscoelasticity on the shear failure of adhesives, showcasing Aluminium acetylacetonate′s role in enhancing the performance and durability of industrial adhesives (Arrowood et al., 2023).
  • UV/Blue Light-Emitting Devices: Highlights the synthesis of UV/blue light-emitting materials using Aluminium acetylacetonate, emphasizing its application in the development of advanced light-emitting diodes (LEDs), contributing to the progress in display and lighting technologies (Lee et al., 2022).
  • Carbon Nanowall Synthesis: Details the use of Aluminium acetylacetonate in the synthesis of carbon nanowalls from a single-source metal-organic precursor, illustrating its effectiveness in nanotechnology and materials science, particularly for enhancing electronic and thermal properties of materials (Giese et al., 2018).

Analysis Note

Assay (out of Al): ≥ 98,0 %(m)
Melting range (lower value): ≥ 192 °C
Melting range (upper value): ≤ 195 °C
Identity (IR): conforms

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.024856747538.14056
form

powder

form

powder

form

powder, solid

form

powder

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

mp

190-193 °C

mp

213-216 °C

mp

190-193 °C (lit.)

mp

260 °C (External MSDS)

bp

315 °C/1013 hPa

bp

-

bp

315 °C (lit.)

bp

-

potency

48.7 mg/kg LD50, oral (Rat)

potency

3360 mg/kg LD50, oral (Rat), 6350 mg/kg LD50, skin (Rabbit)

potency

-

potency

-

pH

8 (20 °C, 3 g/L in H2O)

pH

6 (20 °C, 1 g/L in H2O)

pH

-

pH

-


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

244.4 °F

flash_point_c

118 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters