Skip to Content
Merck
All Photos(1)

Key Documents

8.00553

Sigma-Aldrich

Trimethyl phosphite

for synthesis

Synonym(s):

Trimethyl phosphite

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CZK 946.00
500 ML
CZK 2,640.00

CZK 946.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
CZK 946.00
500 ML
CZK 2,640.00

About This Item

Empirical Formula (Hill Notation):
C3H9O3P
CAS Number:
Molecular Weight:
124.08
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-471-5
NACRES:
NA.22

CZK 946.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

28 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

250 °C

potency

1600 mg/kg LD50, oral (Rat)
2600 mg/kg LD50, skin (Rabbit)

expl. lim.

5.2-61.2 % (v/v)

bp

112 °C/1013 hPa

mp

-78 °C

transition temp

flash point 28 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

P(OC)(OC)OC

InChI

1S/C3H9O3P/c1-4-7(5-2)6-3/h1-3H3

InChI key

CYTQBVOFDCPGCX-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.051 - 1.053
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service