Skip to Content
Merck

8.00258

Allyl cyanide

for synthesis

Synonym(s):

Allyl cyanide, 3-Butenenitrile, Vinylacetonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH2=CHCH2CN
CAS Number:
Molecular Weight:
67.09
UNSPSC Code:
12352117
EC Index Number:
203-701-1
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

form

liquid

autoignition temp.

455 °C

potency

115 mg/kg LD50, oral (Rat), 1170 mg/kg LD50, skin (Rabbit)

mp

-87 °C

transition temp

flash point 23 °C

density

0.83 g/cm3 at 25 °C

storage temp.

2-8°C

InChI

1S/C4H5N/c1-2-3-4-5/h2H,1,3H2

InChI key

SJNALLRHIVGIBI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.830 - 0.835
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Allyl cyanide


pictograms

FlameSkull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

75.2 °F - closed cup

flash_point_c

24 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library