Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
250 mL | Please contact Customer Service for Availability | CZK 1,570.00 CZK 1,177.50 |
About This Item
CZK 1,177.50
List PriceCZK 1,570.00Save 25%vapor pressure
0.003 hPa ( 20 °C)
Quality Segment
assay
≥98.0% (GC)
form
liquid
autoignition temp.
460 °C
potency
138 mg/kg LD50, oral (Rat), 2134 mg/kg LD50, skin (Rabbit)
expl. lim.
7-14 % (v/v)
mp
2 °C
transition temp
flash point 93 °C (decomposition)
solubility
50 g/L
density
0.96 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
N#CCCCCC#N
InChI
1S/C6H8N2/c7-5-3-1-2-4-6-8/h1-4H2
InChI key
BTGRAWJCKBQKAO-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 0.962 - 0.964
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98.0% (GC) | assay 99% | assay >99% | assay - |
| solubility 50 g/L | solubility - | solubility - | solubility - |
| Quality Level 200 | Quality Level 100 | Quality Level 100 | Quality Level 200 |
| mp 2 °C | mp 1-3 °C (lit.) | mp 1-3 °C (lit.) | mp - |
| storage temp. 2-30°C | storage temp. - | storage temp. - | storage temp. - |
| density 0.96 g/cm3 at 20 °C | density 0.951 g/mL at 25 °C (lit.) | density 0.951 g/mL at 25 °C (lit.) | density 1.021 g/mL at 25 °C (lit.) |
Still not finding the right product?
Explore all of our products under Adiponitrile
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation
Storage Class
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 1
flash_point_f
325.4 °F - closed cup
flash_point_c
163 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



