Skip to Content
Merck

Skip To

79145BC

GDS PPMX Microsoft Surface Laptop & Keyboard Cover

Laptop for use with Assurance® GDS PickPen® PIPETMAX® system

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
102 500,00 CZK

About This Item

UNSPSC Code:
41121521
NACRES:
NC.81

102 500,00 CZK


Please contact Customer Service for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For use with GDS PICKPEN™ PIPETMAX™ System

Quality Segment

manufacturer/tradename

Assurance GDS® PickPen, BioControl Systems 79145

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS

storage temp.

room temp

General description

GDS PPMX is supported with the Microsoft Surface Pro Laptop, which work faster and provide flexible, simplified and secure solutions. The Surface Laptop is designed from the inside out to make a bigger impact on productivity, faster multitasking, greater security and connectivity. TRILUTION micro software manages your automated PIPETMAX® workflows, allowing you to focus on more important tasks. The software is easy to learn and allows anyone in the lab to effortlessly manage PIPETMAX® tasks.

Application

GDS PPMX Microsoft Surface Laptop pre-installed with the TRILUTION micro software provides complete automated PIPETMAX® workflows, allowing users to focus on more important tasks.
PCR, Immunomagnetic Separation (IMS)

Features and Benefits

  • Manage protocols, including importing, exporting, and deleting protocols
  • Run pre-loaded Assurance® GDS protocols
  • LIMS compatible

Other Notes

One Microsoft Surface laptop and keyboard cover

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.
PICKPEN is a registered trademark of Merck KGaA, Darmstadt, Germany
PIPETMAX is a registered trademark of Gilson, Inc

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
73071BC79146BC79155BC
description

Laptop for use with Assurance® GDS PickPen® PIPETMAX® system

description

Laptop for use with Assurance® GDS Rotor-Gene® Q thermocycler

description

BioControl Systems, for use with GDS PPMX Microsoft Surface Laptop

description

BioControl Systems, for use with GDS PickPen® PIPETMAX System

compatibility

BioControl GDS

compatibility

for use with GDS Rotor-Gene

compatibility

for use with GDS PPMX Microsoft Surface Laptop

compatibility

for use with GDS PickPen® PIPETMAX System

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

description

For use with GDS PICKPEN™ PIPETMAX™ System

description

-

description

-

description

-

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

application(s)

food and beverages

application(s)

food and beverages
pathogen testing

manufacturer/tradename

Assurance GDS® PickPen, BioControl Systems 79145

manufacturer/tradename

Assurance GDS®, BioControl Systems 73071

manufacturer/tradename

BioControl Systems

manufacturer/tradename

BioControl Systems


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters