Skip to Content
Merck
All Photos(1)

Key Documents

71757-M

Sigma-Aldrich

Overnight Express Instant LB Medium - Novagen

Synonym(s):

Express LB Medium, Instant LB Medium, Novagen LB Medium

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 1,780.00
5 EA
CZK 12,700.00
1 KG
CZK 18,500.00

CZK 1,780.00


Estimated to ship onMay 05, 2025Details



Select a Size

Change View
1 EA
CZK 1,780.00
5 EA
CZK 12,700.00
1 KG
CZK 18,500.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.54

CZK 1,780.00


Estimated to ship onMay 05, 2025Details


form

solid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

technique(s)

microbiological culture: suitable

application(s)

microbiology

storage temp.

10-30°C

General description

Overnight Express Instant LB Medium is a granulated autoinduction culture medium for high-level protein production in pET and other IPTG-inducible bacterial expression systems. Instant LB Medium is a combination of Overnight Express Autoinduction System 1 components (Cat. No. 71300) with Luria Bertani (LB) broth and is supplied in two formats. EasyPak is an aluminum foil pouch containing 45 g granulated medium, sufficient for 1 L culture. Just add the EasyPak contents to 1 L sterile water, supplement with 10 ml glycerol, and microwave for 2 min to dissolve.

Overnight Express Instant LB Medium is also available in 1 kg bottles. Granulated medium ensures rapid, uniform dissolution in water, prevents clumping of the medium, and avoids inhalation of airborne powder. Overnight Express autoinduction media are based on technology developed by F. William Studier at Brookhaven National Laboratory and enable regulated protein expression in E. coli, without monitoring the culture or adding IPTG during cell growth. The method uses media components that are metabolized differentially to promote growth to high cell density and automatically induce protein expression from lac promoters. As a result, these unique autoinduction media provide high-level protein production in pET and other IPTG-inducible bacterial expression systems. Cell mass and target protein yield are often increased several-fold compared to conventional protocols using induction with IPTG. Autoinduction media are extremely convenient for routine expression of proteins in multiple cultures and are ideal for high-throughput parallel analysis of protein expression, solubility, and purification from multiple expression clones.
PRODUCT USE RESTRICTIONS - PRODUCT LABEL LICENSE

The Overnight Express Auto Induction Media material (Products) includes patented technology embodied therein developed at Brookhaven National Laboratory under contract with the U.S. Department of Energy. The patent applications or issued patents embodied in this Product were assigned to Brookhaven Science Associates, LLC (BSA). BSA has enabled EMD Millipore and its Affiliates to grant a non-exclusive license for use of this patented technology, including use with the enclosed Products, based upon your acceptance of the following terms:

1. These Products are to be used for non-commercial research purposes only. For the avoidance of doubt, such non-commercial research purposes include research use by academic, non-profit, and commercial entities.

2. A separate license is required for any commercial use, including the use of the enclosed Products for developing or making other materials for sale to third parties.

3. Information about licenses for commercial use of the Products may be obtained from:
Office of Technology Commercialization and Partnerships
Brookhaven National Laboratory
Building No. 490C - P.O. Box 5000
Upton, New York 11973-5000 Telephone: 631-344-4711 Fax: 631-344-3729
E-mail: [email protected]

4. Information about licenses for commercial manufacture and/or sale of the Products may be obtained from:
Intellectual Property Department
EMD Millipore Corporation
28820 Single Oak Drive
Temecula, CA 92590
E-mail: [email protected]

5. No Products that contain this patented technology may be transferred or distributed further to third parties outside of your laboratory, unless the recipient third party receives a license from BSA and agrees to be bound by its terms.

By keeping or using the enclosed Product(s), you accept the above terms of use solely for noncommercial research purpose. If you refuse to accept the terms herein, you must return the Product(s) to EMD Millipore.

Warning

Toxicity: Standard Handling (A)

Legal Information

Express is a trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service