Skip to Content
Merck
All Photos(1)

Key Documents

71344-M

Sigma-Aldrich

Origami 2 Competent Cells - Novagen

Escherichia coli, rod shaped

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106202
NACRES:
NA.81
Product 71344-M is not currently sold in your country. Contact Technical Service

Product Name

Origami 2 Competent Cells - Novagen, Origami 2 strains have mutations in glutathione reducatase (gor) and thioredoxin reductase (trxB), facilitating proper disulfide bond formation.

biological source

Escherichia coli

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

growth mode

adherent or suspension

morphology

rod shaped

technique(s)

microbiological culture: suitable

cell transformation

transformation efficiency: 2×106 cfu/μg

shipped in

dry ice

storage temp.

−70°C

General description

Genotype: Δ(ara-leu)7697 ΔlacX74 ΔphoA PvuII phoR araD139ahpC galE galK rpsL F′[lac+ lacIq pro] gor522::Tn10trxB (StrR, TetR)

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges us to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.
Origami 2 strains have mutations in glutathione reducatase (gor) and thioredoxin reductase (trxB), facilitating proper disulfide bond formation.
Origami 2 host strains are K-12derivatives that have mutations in both thethioredoxin reductase (trxB) and glutathionereductase (gor) genes, which greatly enhancedisulfide bond formation in the E. colicytoplasm. The Origami2 strains are kanamycin sensitive; like theoriginal Origami strains, the gor mutation is still selectedfor by tetracycline. To reduce the possibility ofdisulfide bond formation between molecules,strains containing mutations in trxB and gor arerecommended only for the expression of proteinsthat require disulfide bond formation for properfolding.

Components

0.4 ml1 mlComponent

•2 × 0.2 ml5 × 0.2 mlOrigami 2 Competent Cells

•2 × 2 ml4 × 2 mlSOC Medium

•2 ng2 ngTest Plasmid

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service