Skip to Content
Merck
All Photos(1)

Key Documents

71337-M

Sigma-Aldrich

pCDF-2 Ek/LIC Vector Kit - Novagen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106602
NACRES:
NA.54
Product 71337-M is not currently sold in your country. Contact Technical Service

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

storage temp.

−70°C

General description

The pCDF-2 Ek/LIC Vector is a LIC-compatible derivative of pCDF-1b. The plasmid contains a strong T7lac promoter and an amino terminal His•Tag coding sequence immediately followed by an Ek/LIC cloning site designed to allow the generation of fusion proteins with minimal vector-encoded sequence. An optional C-terminal S•Tag coding sequence is compatible with purification, detection, and quantification (1). This plasmid is compatible with the LIC Duet Adaptors for coexpression of two proteins as well as with pET-46 and pRSF-2 Ek/LIC vectors for coexpression of up to six proteins when using the LIC Duet Adaptors.




Commercial use of this Product requires a commercial license from EMD Millipore Corporation. Commercial use shall include but not be limited to (1) use of the Product or its components in manufacturing; (2) use of the Product or its components to provide a service, information, or data to others in exchange for consideration; (3) use of the Product or its components (or any derivatives thereof) for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the Product or its components, whether or not such Product or its components are resold for use in research. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of this Product.

Components

•: 1 µg: Ek/LIC Vector


•: 25 units: T4 DNA Polymerase, LIC-qualified


•: 50 µl: 25 mM EDTA


•: 40 µl: 25 mM dATP


•: 8 µl: Ek/LIC β-gal Control Insert


•: 100 µl: 100 mM DTT


•: 50 µl: 10X T4 DNA Polymerase Buffer


•: 1.5 ml: Nuclease-free Water


•: 5 × 2 ml: SOC Medium


•: 10 µl: Test Plasmid


•: 0.2 ml: BL21(DE3)pLysS Competent Cells


•: 0.2 ml: BL21(DE3) Competent Cells


•: 22 × 50 µl: NovaBlue GigaSingles Competent Cells

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Other Notes

1. Kim, J.S. and Raines, R.T. (1993) Protein Science2, 348–356.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service