Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Estimated to ship onJune 04, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 7,960.00 |
About This Item
NACRES:
NA.51
UNSPSC Code:
41106610
CZK 7,960.00
Estimated to ship onJune 04, 2026Details
form
solid
Quality Segment
manufacturer/tradename
Novagen®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles
storage temp.
−20°C
General description
pSTBlue-1 is a multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, dual opposed T7/SP6 promoters and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3′ overhangs are conveniently positioned near each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the “EMD Product”) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: [email protected].
This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the “EMD Product”) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: [email protected].
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form liquid | form liquid | form liquid |
| storage temp. −20°C | storage temp. −70°C | storage temp. −70°C | storage temp. −20°C |
| manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Novagen® | manufacturer/tradename Novagen® |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage condition OK to freeze | storage condition OK to freeze, avoid repeated freeze/thaw cycles | storage condition OK to freeze | storage condition OK to freeze, avoid repeated freeze/thaw cycles |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



