Skip to Content
Merck
All Photos(1)

Documents

70036-M

Sigma-Aldrich

X-Gluc Solution - Novagen

A colorimetric stain for GUS activity

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

Quality Level

form

solution

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

The β-glucuronidase substrate X-Gluc (5-bromo-4-chloro-3-indolyl-β-D-glucuronide) is a colorimetric stain for GUS activity, and is used to identify recombinants containing pBACgus vectors, and LIC GUS Control Insert. X-Gluc Solution is provided as a 20 mg/ml stock solution, ready for dilution and use in staining plaques for GUS activity. Each vial contains enough substrate to stain 15 100-mm plates.

Warning

Toxicity: Irritant (B)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (refers to pure substance)

Flash Point(C)

87 °C - closed cup - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service