Skip to Content
Merck

Skip To

6505-OP

Phosphate Buffered Saline

10X, liquid, OmniPur® Grade

Synonym(s):

PBS, Phosphate Buffered Saline, 10X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
4 L

Estimated to ship onJuly 22, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 5,450.00
20 L

Estimated to ship onJune 10, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 15,700.00

About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

CZK 5,450.00


Estimated to ship onJuly 22, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphate Buffered Saline, OmniPur® Grade, 10X Liquid Concentrate

Quality Segment

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

ambient

General description

Each liter makes 10 L of 1X PBS (137 mM NaCl, 2.7 mM KCl, 10 mM Phosphate Buffer). Molecular Biology Grade.

DNAse: none detected
pH (10%, water) at 25°C: 7.3-7.5
RNAse: none detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • Maternal Obesity and Kawasaki Disease-like Vasculitis: A New Perspective on Cardiovascular Injury and Inflammatory Response in Offspring Male Mice.: This study utilized phosphate buffered saline (PBS) to explore how maternal obesity affects cardiovascular health and inflammatory responses in offspring, providing insights into potential therapeutic interventions (Zheng et al., 2023).

  • Immunohistochemistry of Leukemia Inhibitory Factor and Integrin αVβ3 in Mouse Endometrium Following Kisspeptin-54 Ovulation Trigger.: PBS was employed for immunohistochemical staining to study the role of Leukemia Inhibitory Factor and Integrin in mouse endometrium, contributing to our understanding of reproductive biology and potential fertility treatments (Abdelkareem et al., 2023).

  • The mechanism of acid resistance by ornithine decarboxylase in Trichinella spiralis.: This investigation utilized PBS to understand the biochemical pathways involved in acid resistance in Trichinella spiralis, which has implications for parasitology and disease control (Wang et al., 2023).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
6506-OP8410-OP8310-OP
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

color

clear colorless

color

clear colorless

color

clear colorless

color

clear colorless

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®


Still not finding the right product?

Explore all of our products under Phosphate Buffered Saline


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. what is the recommended expiration date of this solution once opened?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one-year warranty from the date of shipment.

      For more information, you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters