Skip to Content
Merck
All Photos(1)

Documents

5920-OP

Millipore

Lithium Chloride

OmniPur® Grade, 8M

Synonym(s):

LiCl Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352101
NACRES:
NA.28

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

color

clear colorless

shipped in

ambient

General description

Used for precipitation of RNA prior to PCR or cDNA synthesis. Separates RNA from inhibitors of cDNA synthesis. The 0.2 µm filtered solution is clean, convenient and ready to use.

Specifications:
DNase: None detected
Concentration (molarity): 7.7-8.2
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application


  • LiCl in situ decorated metal-organic framework (MOF)-derived porous carbon for efficient solar-driven atmospheric water harvesting.: This study explores the application of Lithium Chloride (LiCl) in enhancing the efficiency of atmospheric water harvesting using MOF-derived porous carbon materials. The integration of LiCl significantly improves water adsorption and desorption properties, making it a promising material for sustainable water resources (Guo S et al., 2024) (Guo S et al., 2024).

  • WDR1 promotes prostate cancer progression through Wnt/ß-catenin signaling.: Although this study primarily focuses on WDR1′s role in cancer, the use of Lithium Chloride in Wnt signaling pathway modulation offers insights into its potential therapeutic applications in cancer treatment (Cheng J et al., 2024) (Cheng J et al., 2024).

  • Solar-Driven Drum-Type Atmospheric Water Harvester Based on Bio-Based Gels with Fast Adsorption/Desorption Kinetics.: This research presents a bio-based gel system enhanced with Lithium Chloride for rapid water adsorption and desorption, showcasing its potential in efficient water harvesting technologies (Zhou H et al., 2024) (Zhou H et al., 2024).

  • Na,K-ATPase activity promotes macropinocytosis in colon cancer via Wnt signaling.: This article highlights the involvement of Lithium Chloride in Na,K-ATPase activity and Wnt signaling, suggesting its role in cellular processes and potential cancer therapies (Tejeda-Muñoz N et al., 2024) (Tejeda-Muñoz N et al., 2024).

  • PRMT5-mediated arginine methylation of FXR1 is essential for RNA binding in cancer cells.: The study investigates the biochemical pathways involving Lithium Chloride, emphasizing its significance in RNA-binding protein modifications and cancer cell dynamics (Vijayakumar A et al., 2024) (Vijayakumar A et al., 2024).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service