Skip to Content
Merck
All Photos(1)

Key Documents

586005

Millipore

Thapsigargin

≥97% (TLC), solid, endoplasmic reticular Ca2+-ATPase inhibitor, Calbiochem®

Synonym(s):

Thapsigargin - CAS 67526-95-8 - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H50O12
CAS Number:
Molecular Weight:
650.75
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Thapsigargin - CAS 67526-95-8 - Calbiochem, Thapsigargin, CAS 67526-95-8, is a cell-permeable, tumor-promoting sesquiterpene lactone that releases calcium by non-competitvley inhibiting endoplasmic reticular Ca2+-ATPase (IC₅₀ = 4-13 nM).

Quality Level

Assay

≥97% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

solubility

DMSO: >1 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O1[C@@H]2[C@]([C@H](C[C@@]([C@H]3[C@@H]([C@H](C(=C32)C)OC(=O)\C(=C/C)\C)OC(=O)CCCCCCC)(OC(=O)C)C)OC(=O)CCC)([C@@](C1=O)(O)C)O

InChI

1S/C34H50O12/c1-9-12-13-14-15-17-24(37)43-28-26-25(20(5)27(28)44-30(38)19(4)11-3)29-34(41,33(8,40)31(39)45-29)22(42-23(36)16-10-2)18-32(26,7)46-21(6)35/h11,22,26-29,40-41H,9-10,12-18H2,1-8H3/b19-11-/t22-,26+,27-,28-,29-,32-,33+,34+/m0/s1

InChI key

IXFPJGBNCFXKPI-FSIHEZPISA-N

General description

A cell-permeable tumor-promoting sesquiterpene lactone that releases Ca2+ by inhibiting endoplasmic reticular Ca2+-ATPase (IC50 = 4-13 nM). Does not increase inositol phosphates and has no significant effect on protein kinase C. Increases Ca2+-dependent Na+ influx in human platelets in a dose-dependent manner. Induces apoptosis in rat thymocytes and in human hepatoma cells. Shown to induce apoptosis in alveolar rhabdomyosarcoma cells by elevating Akt-stimulated PAX3-FOXO1 phosphorylation and promoting its proteasomal degradation.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
endoplasmic reticular Ca2+ ATPase
Product does not compete with ATP.
Reversible: no
Target IC50: 4-13 nM against endoplasmic reticular Ca2+-ATPase

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for 2-3 months at -20°C.

Other Notes

Jothi, M., et al. 2013. Mol. Cancer Ther. In press.
Won, J.G. and Orth, D.N. 1995. Endocrinology 136, 5399.
Tepel, M., et al. 1994. Biochim. Biophys. Acta1220, 248.
Begum, N., et al. 1993. J. Biol. Chem. 268, 3352.
Low, A.M., et al. 1993. Eur. J. Pharmacol.230, 53.
Tsukamoto, A. and Karneko, Y. 1993. Cell. Biol. Int.17, 969.
Wong, W.L., et al. 1993. Biochem. J.289, 71.
Glennon, M.C., et al. 1992. J. Biol. Chem. 267, 25568.
Xu, Y., et al. 1992. J. Neurochem.59, 2224.
Mason, M.J., et al. 1991. J. Biol. Chem. 266, 20856.
Thastrup, O., et al. 1990. Proc. Natl. Acad. Sci. USA87, 2466.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Birk et al.
Bio-protocol, 11(13), e4076-e4076 (2021-07-31)
The production of reactive oxygen species (ROS) and endoplasmic reticulum (ER) stress are tightly linked. The generation of ROS can be both the cause and a consequence of ER stress pathways, and an increasing number of human diseases are characterized
Cédric Montigny et al.
Scientific reports, 11(1), 1641-1641 (2021-01-17)
Sarcolipin (SLN), a single-spanning membrane protein, is a regulator of the sarco-endoplasmic reticulum Ca2+-ATPase (SERCA1a). Chemically synthesized SLN, palmitoylated or not (pSLN or SLN), and recombinant wild-type rabbit SERCA1a expressed in S. cerevisiae design experimental conditions that provide a deeper
Amena BenYounès et al.
Autophagy, 7(8), 883-891 (2011-04-05)
Autophagic flux can be measured by determining the declining abundance of autophagic substrates such as sequestosome 1 (SQSTM1, better known as p62), which is sequestered in autophagosomes upon its direct interaction with LC3. However, the total amount of p62 results
Tess Holling et al.
Human mutation, 43(5), 625-642 (2022-03-11)
BNIP1 (BCL2 interacting protein 1) is a soluble N-ethylmaleimide-sensitive factor-attachment protein receptor involved in ER membrane fusion. We identified the homozygous BNIP1 intronic variant c.84+3A>T in the apparently unrelated patients 1 and 2 with disproportionate short stature. Radiographs showed abnormalities
A Umano et al.
The Journal of biological chemistry, 298(5), 101899-101899 (2022-04-11)
The spinocerebellar ataxias (SCAs) are a class of incurable diseases characterized by degeneration of the cerebellum that results in movement disorder. Recently, a new heritable form of SCA, spinocerebellar ataxia type 48 (SCA48), was attributed to dominant mutations in STIP1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service