Skip to Content
Merck

Skip To

580563

TCPOBOP

The most potent known member of the phenobarbital-like class of cytochrome P450 (CYP)-inducing agents.

Synonym(s):

TCPOBOP, 1,4-Bis[2-(3,5-dichloropyridyloxy)]benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg

Available to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

CZK 4,380.00

About This Item

Empirical Formula (Hill Notation):
C16H8Cl4N2O2
CAS Number:
Molecular Weight:
402.06
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

CZK 4,380.00


Available to ship TODAYDetails


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - UT7145000

Quality Segment

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Clc1c(ncc(c1)Cl)Oc2ccc(cc2)Oc3ncc(cc3Cl)Cl

InChI

1S/C16H8Cl4N2O2/c17-9-5-13(19)15(21-7-9)23-11-1-2-12(4-3-11)24-16-14(20)6-10(18)8-22-16/h1-8H

InChI key

BAFKRPOFIYPKBQ-UHFFFAOYSA-N

General description

A highly potent inducer of phenobarbital-like class of cytochrome P450 (CYP) enzymes. A tumor promoter that induces hepatocyte proliferation, independent of the presence of cytokines or transcription factors. Activates orphan nuclear receptors mCAR (mouse constitutive androstane receptor) and hPXR (human pregane X receptor), but has little or no effect on hCAR and mPXR. Also suppresses the expression of multi-drug resistance genes mdr2 and mdr3 in mouse liver.
The most potent known member of the phenobarbital-like class of cytochrome P450 (CYP)-inducing agents. Acts as a tumor promoter of murine hepatocarcinogenesis; activates orphan nuclear receptors mCAR (mouse Constitutive Androstane Receptor) and hPXR (human Pregnane X Receptor), but has little or no activity on hCAR and mPXR. Also suppresses the multidrug resistance gene expression in the mouse liver.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Cytochrome P450 (CYP)-inducing agents
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ledda-Columbano, G.M., et al. 2000. Am. J. Pathol.156, 91.
Moore, L.B., et al. 2000. J. Biol. Chem.275, 15122.
Tzameli, I., et al. 2000. Mol. Cell. Biol.20, 2951.
Forman, B.M., et al. 1998. Nature395, 612.
Honkakoski, P., et al. 1998. Mol. Pharmacol.53, 597.
Russell, A.L., et al. 1994. Int. J. Cancer58, 550.
Smith, G., et al. 1993. Biochem. J.289, 807.
Dargani, T.A., et al. 1990. Carcinogenesis11, 1153.
Poland, A., et al. 1980. Mol. Pharmacol.18, 571.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T1443219491524400
assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

form

solid

form

solid

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 5 mg/mL

solubility

DMSO: >10 mg/mL, H2O: insoluble

solubility

DMSO: 5 mg/mL

solubility

DMSO: 10 mg/mL


Still not finding the right product?

Explore all of our products under TCPOBOP


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters