Skip to Content
Merck

Skip To

567569

Spautin-1

a quinazolin compound that acts as a specific and potent inhibitor of autophagy

Synonym(s):

Spautin-1, Autophagy Inhibitor II, C43, 6-fluoro-N-[4-fluorobenzyl]quinazolin-4-amine, Specific and Potent AUtophagy Inhibitor 1, C43, 6-fluoro-N-[4-fluorobenzyl]quinazolin-4-amine, Specific and Potent AUtophagy Inhibitor 1, Autophagy Inhibitor II

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg

Estimated to ship onJune 01, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 5,030.00

About This Item

Empirical Formula (Hill Notation):
C15H11F2N3
Molecular Weight:
271.26
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥99% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light

CZK 5,030.00


Estimated to ship onJune 01, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)CNc2ncnc3c2cc(cc3)F

InChI

1S/C15H11F2N3/c16-11-3-1-10(2-4-11)8-18-15-13-7-12(17)5-6-14(13)19-9-20-15/h1-7,9H,8H2,(H,18,19,20)

InChI key

AWIVHRPYFSSVOG-UHFFFAOYSA-N

General description

A quinazolin compound that acts as a specific and potent inhibitor of autophagy and promotes Vps34 PI 3-kinase complex degradation by blocking the activity of USP 10 (IC50 = 580 nM) and USP 13 (IC50 = 690 nM) deubiquitinating enzymes. However, it does not affect the lipid kinase activity of Vps34. Also shown to reduce the level of cytoplasmic and nuclear p53. Sensitizes breast cancer cell lines (BCAP37, MCF-7, and BT549) to apoptosis under conditions of nutritional deprivation. Also shown to inhibit etoposide-induced cell death in Bax/Bak double knockout (DKO) cells. Normal cell lines (e.g., MDCK cells) do not undergo autophagy in the presence of Spautin-1.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lui, J., et al. 2011, Cell.147, 223.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
260964681669239804
assay

≥99% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥70% (sum of two isomers, HPLC)

form

solid

form

solid

form

solid

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light


pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters