Skip to Content
Merck
All Photos(1)

Key Documents

567538

Sigma-Aldrich

Sodium Nitroprusside, Dihydrate

Nitric oxide donor. Reported to reduce lipid peroxidation-induced membrane damage.

Synonym(s):

Sodium Nitroprusside, Dihydrate, SNP, Sodium Nitroferricyanide(III) Dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 1,670.00

CZK 1,670.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 MG
CZK 1,670.00

About This Item

Linear Formula:
Na2[Fe(CN)5]NO · 2H2O
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

CZK 1,670.00


In StockDetails


Request a Bulk Order

Quality Level

Assay

>99% (titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

red

solubility

water: soluble

shipped in

ambient

storage temp.

10-30°C

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;-2;;2*+1;;

InChI key

NWFKTGARQFKZMN-UHFFFAOYSA-N

General description

Nitric oxide donor. Reported to reduce lipid peroxidation-induced membrane damage.

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, store at room temperature (20°C). Stock solutions are stable for up to 2 weeks at 20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Hellstrom, W.J., et al. 1994. Fertil. Steril. 61, 1117.
Kowaluk, E.A., et al. 1992. J. Pharmacol. Exp. Ther.262, 916.
Ignarro, L.J., et al. 1981. J. Pharmacol. Exp. Ther.218, 739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service