Skip to Content
Merck

Skip To

565000

Serine Protease Inhibitor Cocktail Set I - Calbiochem

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 vial

Estimated to ship TODAYfromAreál Kühne+Nagel spol. s r.o.

2 530,00 CZK
5 vials

Estimated to ship on11 September 2026fromAreál Kühne+Nagel spol. s r.o.

11 000,00 CZK

About This Item

NACRES:
NA.28
UNSPSC Code:
12352209

2 530,00 CZK


Estimated to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

manufacturer/tradename

(Calbiochem)

storage condition

dry at room temperature (Tightly closed)

technique(s)

inhibition assay: suitable

input

general use sample(s)

solubility

water: soluble

suitability

suitable for activity/function inhibition

UniProt accession no.

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

-10 to -25°C

Gene Information

human ... SERPINA(5104)

General description

The Serine Protease Inhibitor Cocktail Set I, called Serpins, is a mixture of highly specific inhibitor protein that provides a broad range of serine protease inhibition. This product specifically inhibits serine proteases using inhibitors such as 4-benzenesulfonyl fluoride hydrochloride (AEBSF), and Aprotinin.

Application

The Serine Protease Inhibitor Cocktail Set I – Calbiochem may be used:
  • to protect proteins from serine protease degradation during cell or tissue lysis to preserve sample integrity.
  • to maintain protein stability and native state in downstream proteomic analyses such as Western blotting, immunoprecipitation, and mass spectrometry.
  • to support accurate protein quantification and characterization in studies involving biomarker discovery and post-translational modifications.

Biochem/physiol Actions

Serine proteases are enzymes involved in processes like coagulation, fibrinolysis, inflammation, apoptosis, and tissue homeostasis. It acts through complimentary and overlapping inhibitory mechanisms. Serpins like 4-Benzenesulfonyl fluoride hydrochloride (AEBSF) inhibit serine proteases by reacting rapidly with the enzyme′s catalytic serine residue. This reaction forms a stable acyl-enzyme complex that prevents protease from breaking down proteins to stop its activity.


Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library