Skip to Content
Merck
All Photos(2)

Key Documents

539124

Millipore

Protease Arrest Reagent

An optimized concentration of various reversible and irreversible inhibitors to inhibit serine, cysteine, and calpain proteases.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

storage temp.

2-8°C

General description

An optimized concentration of various reversible and irreversible inhibitors to inhibit serine, cysteine, and calpain proteases. Suitable for the protection of proteins purified from animal tissues, plant tissues, yeast, and bacteria. EDTA is also provided separately to inhibit metalloproteinases. Protease Arrest Reagent is provided as a 100X solution that when diluted in extraction buffer at pH 7.0-8.0 inhibits 95-98% of protease activity. Supplied with a data sheet.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Serine, cysteine, and calpain proteases

Warning

Toxicity: Irritant (B)

Reconstitution

Following addition to buffers or extracts this product is stable for up to 2 weeks at 4°C or for up to 3 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service