Skip to Content
Merck
All Photos(1)

Key Documents

5.38001

Millipore

T-00127-HEV1

Synonym(s):

T-00127-HEV1, 3-(3,4-Dimethoxyphenyl)-2,5-dimethyl-N-(2-morpholinoethyl)pyrazolo[1,5-a]pyrimidin-7-amine, PI4KIIIβ Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29N5O3
CAS Number:
Molecular Weight:
411.50
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyrazolopyrimidinamine derived enviroxime-like compound that displays anti-poliovirus activity with broad specificity for enteroviruses (EC50 = 0.77, 3.38, 2.5, 1.03 & >44 µM for PV, CVB3, HRVM, HCV 1b & HCV 2a, respectively). Dose-dependently inhibits viral RNA replication. Acts as a potent, selective, ATP-competitive and reversible inhibitor of PI4KIIIβ (IC50 = 150 nM) with poor affinity towards PI4KIIIα & PI4KIIα (IC50 = ~75 & ≥100 µM). Shown to cause PI4KIIIβ accumulation and reduce PI4P at the Golgi apparatus. Exhibits low cytotoxicity (CC50 >50 µM in HeLa cells) and attractive PK profile in mice. PI4KIIIβ Inhibitor I, BF738735 (Cat. No. 533657) is also available.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PI4KIIIβ
Reversible: yes
Target IC50: 150 nM for PI4KIII&beta

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20&degC). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Mejdrova, I., et al. 2015. J. Med. Chem.58, In press.

Spickler, C., et al. 2013. Antimicrob. Agents Chemother.57, 3358.
Sasaki, J., et al. 2012. EMBO J.31, 754.
Arita, M., et al. 2011. J. Virol.85, 2364.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service