Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C31H31NO3
CAS Number:
Molecular Weight:
465.58
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
assay
≥98% (HPLC)
Quality Segment
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
white
solubility
DMSO: 8 mg/mL
storage temp.
−20°C
General description
A cell-permeable, bioavailable, tetrahydroisoquinolin amide that acts as a selective, potent, and high-affinity agonist of REV-ERB (IC50 = 170 nM for REV-ERBα and IC50 = 160 nM for REV-ERBβ in a REV-ERB ligand-binding domain co-transfection assay). Able to cross the blood brain barrier and remain above the IC50 value even after 6 h (30 mg/kg, i.p.). Displays minimal activities against a wide of other receptors, ion channels, and transporters when screened at 20 µM. Shown to induce wakefulness and suppress circadian wheel-running activity (ED50 = 20 mg/kg injected at Zeitgeiber time 6 (ZT6)) in mice. Reduces slow-wave sleep (SWS) and REM sleep when injected at ZT6 with decreased duration and reduces the number of episodes and duration of REM sleep. Also shown to reduce anxiety-like behavior in the marble-burying assay.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
REV-ERB
REV-ERB
Reversible: yes
Target IC50: 170 nM for REV-ERB&alpha
Packaging
Packaged under inert gas
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Banerjee, S., et al. 2014. Nat. Comm.5, 5759.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.