Skip to Content
Merck

Skip To

5.33871

UniPR129

≥98% (HPLC), Ephrin recptor antagonist, solid

Synonym(s):

Eph Receptor Antagonist, UniPR129, (S)-3-((R)-4-((3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)-4-(1H-indol-3-yl)butanoic acid, Eph Inhibitor, UniPR129

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C36H52N2O4
Molecular Weight:
576.81
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Eph Receptor Antagonist, UniPR129,

assay

≥98% (HPLC)

Quality Segment

form

solid

potency

945 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable L-homo-Trp conjugate of lithocholic acid that acts as an Eph receptor antagonist and competitively disrupts Eph2A-ephrin-A1 binding (IC50 = 945 nM; KI = 345 nM). Blocks the interaction of ephrin-A1 and ephrin-B1 with all Eph receptors (IC50 = 840 nM - 1.54 µM for EphA and 2.6 - 3.74 µM for EphB). Shown to inhibit EphA2 phosphorylation in PC3 prostate cancer cells (IC50 = 5 µM), however, it does not affect tyrosine kinase activity of EGF and VEGF receptors. Slightly reduces PC3 cell proliferation without affecting cell cycle or apoptosis. However, it blocks the rounding effect of ephrin-A1-Fc in PC3 cells (IC50 = 6.25 µM). Also shown to inhibit angiogenesis in HUVEC (IC50 = 5.2 µM) and reduce ephrin-induced activation of EPhA2 and EphB4 (IC50 = 26.3 and 18.4 µM, respectively).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Eph Receptors
Reversible: yes

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hassan-Mohamed, I., et al. 2014. Brit. Journ. Pharm.171, 5195.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters