Skip to Content
Merck

5.33869

Sigma-2 Receptor Antagonist, CT01344

Synonym(s):

Sigma-2 Receptor Antagonist, CT01344, (R)-4-(3,4-dichlorophenyl)-N-(4-(trifluoromethyl)benzyl)butan-2-amine, PGRMC-1 Receptor Antagonist, CT01344

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H18Cl2F3N
CAS Number:
Molecular Weight:
376.24
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
oil
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

Quality Segment

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

light yellow

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable, benzyl amine compound that can cross the blood-brain barrier and act as a selective antagonist of Sigma-2/progesterone receptor membrane component 1 (PGRMC1) receptor. Shown to competitively displace selective radioligand (3H-DTG) from homogenates of human B cell lines (Ki = 48 nM). Effectively blocks the effects of Abeta on membrane trafficking (EC50 = 8.7 µM) and displaces Abeta binding on cultured neurons (EC50 = 3.9 µM). It can also reverse Abeta oligomer-mediated trafficking deficits (EC50 = 7.3 µM) and restore normal synaptic function and cognitive abilities in a murine model of Alzheimer′s disease (10 - 30 mg/kg. p.o. over a period of 5.5 months). Shown to be effective when added either 1 hour prior to or 1 hour after Abeta oligomer preparations.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Primary Target
Sigma-2

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Izzo, P.M., et al. 2014. Plos One.9, e111898.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library