Skip to Content
Merck
All Photos(1)

Key Documents

5.32310

Sigma-Aldrich

c-Myc Inhibitor IV, KJ-Pyr-9

Synonym(s):

c-Myc Inhibitor IV, KJ-Pyr-9, 4-(2-(Furan-2-yl)-6-(4-nitrophenyl)pyridin-4-yl)benzamide, KJPyr9, Krohnke Pyridine-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H15N3O4
CAS Number:
Molecular Weight:
385.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)c2nc(cc(c2)c4ccc(cc4)C(=O)N)c3[o]ccc3

InChI

1S/C22H15N3O4/c23-22(26)16-5-3-14(4-6-16)17-12-19(15-7-9-18(10-8-15)25(27)28)24-20(13-17)21-2-1-11-29-21/h1-13H,(H2,23,26)

InChI key

GTTDVYCKFQYVNN-UHFFFAOYSA-N

General description

A cell-permeable, blood-brain barrier permeant, bioavailable trisubstituted pyridine compound that inhibits c-Myc transcriptional activity. Shown to directly bind to both the monomeric c-Myc (Kd = 6.5 nM) and to c-Myc-Max heterodimer (Kd = 13.4 nM) with high affinity and disrupt c-Myc-Max interaction. Displays a very weak affinity towards Max-Max homodimer (Kd >1 µM). Also interferes with Max homodimerization, albeit to a lesser extent than c-Myc-Max heterodimerization. Shown to inhibit the c-Myc-driven proliferation of NCI-H460, MDA-MB-231, and SUM-159PT and several other cancer cell lines (IC50 = 5-10 µM). Also diminishes the proliferation of Burkitt lymphoma cell lines expressing high levels of c-Myc (IC50 = 2.5 µM). Suppresses the growth of MDA-MB-231 cells in a xenografted nude mice model (10 mg/kg, i.p., q.d.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable, trisubstituted pyridine compound that displays Myc-selective affinity (Kd = 6.5 nM/Myc homodimer, 13.4 nM/Myc-Max dimer, >1.0 µM/Max homodimer) and is 4-times more potent against Myc-Max than Max-Max in DNA-binding assays. Effectively prevents focal microtumors formation following N-Myc, c-Myc, as well as ATG- or CAG-c-Myc transformation of cultured chick embryo fibroblasts/CEF (>99.9% inhibition at 10 µM in ATG-c-Myc-transformed cultures), while exhibiting much reduced or little potency against cultures transformed by v-Src (no inhibition up to 20 µM), v-Jun or PI 3-K H1047R (45.5% inhibition at 10 µM). Selectively inhibits Myc-dependent proliferation of human & avian cultures (Effective conc. 25 to 50 µM; IC50 1 to 10 µM), while exhibiting little potency against Myc-independent growths of human skin fibroblasts, non-transformed, v-jun-transformed, or methylcholanthrene-transformed quail embryo fibroblasts even at a high concentration of 50 µM. Reported to suppress MDA-MB-231-derived tumor expansion in mice (10 mg/kg/d i.p.) in vivo with a concomitant upregulation of N-Myc downregulated gene 1/NDRG1 protein level in tumor tissue. Pharmacokinetics studies reveal good blood-brain barrier permeability in mice ([Drug] = 3.5 µM/plasma & 12.4 µM/brain 4 h post single 10 mg/kg i.p. dosage) and a plasma half-life of 1.84 h in rats following a single i.v. dose of 1 mg/kg.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Myc homodimer
Secondary Target
Myc-Max

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Use only fresh DMSO for reconstitution.

Other Notes

Hart, J.R., et al. 2014. Proc. Natl. Acad. Sci. USA.111, 12556.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service