Skip to Content
Merck
All Photos(1)

Key Documents

5.31661

Sigma-Aldrich

SMYD2 Inhibitor, AZ505

Synonym(s):

SMYD2 Inhibitor, AZ505, N-Cyclohexyl-3-(3,4-dichlorophenethylamino)-N-(2-(2-(5-hydroxy-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-8-yl)ethylamino)ethyl)propanamide, HMTase Inhibitor XV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38Cl2N4O4
CAS Number:
Molecular Weight:
577.54
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C29H38Cl2N4O4/c30-23-8-6-20(18-24(23)31)10-13-32-15-12-27(38)35(22-4-2-1-3-5-22)17-16-33-14-11-21-7-9-25(36)28-29(21)39-19-26(37)34-28/h6-9,18,22,32-33,36H,1-5,10-17,19H2,(H,34,37)

InChI key

LIBVHXXKHSODII-UHFFFAOYSA-N

General description

A cell-permeable benzooxazinyl-ethylaminoethyl-propanamide compound that acts as a potent and reversible inhibitor of SMYD2 (IC50 = 120 nM, Ki = 300 nM, Kd = 500 nM). Shown to bind to the peptide-binding groove of the enzyme. This binding is dependent on the presence of S-adenosylmethionine (SAM). The inhibition appears to be competitive with respect to substrate and uncompetitive with respect to SAM. Displays ~700-fold greater selectivity over SMYD3, DOT1L, EZH2, GLP, G9a and SET7/9 protein lysine methyl transferases (IC50 >83.3 µM). Inhibits SMYD2-mediated p53-K370 methylation in U2OS cells (at ~10 µM), but does not affect methylation in cells transfected with a Y240F mutant.

Please note that the molecular weight for this compound is batch-specific due to variable water content.
AZ505 is a cell-permeable, potent, reversible inhibitor of SMYD2 (IC₅₀ = 120 nM, Ki = 300 nM) that binds to the peptide-binding groove of the enzyme.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
SMYD2

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a di-trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Ferguson, A.D., et al. 2011. Structure19, 1262.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service