Skip to Content
Merck

5.31401

Silicone tubing Ø 6,4 mm

for use with Dilucult, pkg of 5 tubes

Synonym(s):

Dilutor Tubing, Hose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41122409
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

silicone tubing

packaging

pkg of 5 tubes

manufacturer/tradename

DiluCult

application(s)

food and beverages

compatibility

for use with Dilucult

General description

Set of 5 dispensing tubings (silicone tubings + tygon tubings + straight connectors) - inner ø 6.4 mm

Application

For use with DiluCult and DiluCult 2 gravimetric dilutor instrument

Legal Information

DILUCULT is a trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library