Skip to Content
Merck

Skip To

5.30613

IRAP Inhibitor, HFI-419

Synonym(s):

IRAP Inhibitor, HFI-419, (±)-Ethyl-2-acetamido-7-hydroxy-4-(pyridin-3-yl)-4H-chromene-3-carboxylate, HFI419, Insulin-Regulated Aminopeptidase Inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg

Estimated to ship onJuly 30, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 7,020.00

About This Item

Empirical Formula (Hill Notation):
C19H18N2O5
CAS Number:
Molecular Weight:
354.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
PubChem Substance ID:
Assay:
≥97% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light

CZK 7,020.00


Estimated to ship onJuly 30, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (HPLC)

Quality Segment

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CCOC(=O)C1=C(OC2=C(C1C3=CN=CC=C3)C=CC(=C2)O)NC(=O)C

InChI

1S/C19H18N2O5/c1-3-25-19(24)17-16(12-5-4-8-20-10-12)14-7-6-13(23)9-15(14)26-18(17)21-11(2)22/h4-10,16,23H,3H2,1-2H3,(H,21,22)

InChI key

GGBHINRNYAAYMX-UHFFFAOYSA-N

General description

The racemic mixture of an aqueous soluble (at least 50 µg/mL or 140 µM at pH 6.5) pyridinyl-4H-benzopyran compound that inhibits insulin-regulated (IRAP) aminopeptidase activity by competing against substrate for IRAP extracellular catalytic site binding (Ki = 0.48 µM; using 25 µM Leu-AMC as substrate) with concomitant interaction with active site zinc via its acetamide, while displaying little potency against glucose-6-phosphatase, aminopeptidase N, ACE1, leukotriene A4 hydrolase, or ER-associated aminopeptidases 1 & 2 (≤13% inhibition at 100 µM). Reported to boost 1 mM dibutyryl cAMP-evoked glucose uptake in cultured rat hippocampal slices (68% higher uptake in 5 min than dbcAMP alone without 100 nM HFI-419) in vitro and exhibit in vivo memory-enhancing efficacay in an object recognition test when administered directly into the cerebral lateral ventricle of rats via a cannula implant (0.1 or 1.0 nmol/2 µL/rat). HFI-419 is reported to hydrolyze into slightly less potent HFI-142 (Ki = 2.0 µM) after i.v. (plasma t1/2 = 11 min; 2 mg/kg) or i.p (plasma t1/2 = 4.6 h; 10 mg/kg) injection in rats, although HFI-142 is more stable and likely exhibits better blood-brain permeability (Blood:Brain HFI-142 ratio = 3.36 and 0.38, respectively, 0.5 and 4 h post single 3 mg HFI-419/kg i.v. injection).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IRAP
Reversible: yes

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Mountford, S.J., et al. 2014. J. Med. Chem.57, 1368.
Albiston, A.L., et al. 2011. Br. J. Pharmacol.164, 37.
Albiston, A.L., et al. 2010. Mol. Pharmacol.78, 600.
Albiston, A.L., et al. 2008. FASEB J.22, 4209.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
5.33916555562400087
assay

≥97% (HPLC)

assay

≥96% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (sum of two isomers)

form

powder

form

film

form

solid

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters