Skip to Content
Merck
All Photos(1)

Key Documents

529582

Sigma-Aldrich

3-MB-PP1

≥97% (HPLC), solid, PP1 analog, Calbiochem®

Synonym(s):

PP1 Analog III, 3-MB-PP1, 1-(tert-Butyl)-3-(3-methylbenzyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine, Plk1 ASKA Inhibitor, Syk ASKA Inhibitor, ZAP-70 ASKA Inhibitor, Syk ASKA Inhibitor, ZAP-70 ASKA Inhibitor, 1-(tert-Butyl)-3-(3-methylbenzyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine, Plk1 ASKA Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N5
CAS Number:
Molecular Weight:
295.38
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

PP1 Analog III, 3-MB-PP1, PP1 Analog III, 3-MB-PP1 primarily used in Inhibition.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]1(nc(c3c1ncnc3N)Cc2cc(ccc2)C)C(C)(C)C

InChI

1S/C17H21N5/c1-11-6-5-7-12(8-11)9-13-14-15(18)19-10-20-16(14)22(21-13)17(2,3)4/h5-8,10H,9H2,1-4H3,(H2,18,19,20)

InChI key

FYCOTGCSHZKHPR-UHFFFAOYSA-N

General description

A cell-permeable C3-enlarged PP1 (Cat. No. 567809) analog that is shown to act as a potent, ATP-competitive, and highly ASKA- (analog-sensitive kinase alleles) selective inhibitor against polo-like kinase 1C67V L130G (75% and 94% inhibition in cell-free kinase assays at 0.1 and 1 µM, respectively), ZAP-70C405V M414A/ZAP-70M414A (complete prevention and elimination, respectively, of αTCR-stimulated [Ca2+] elevation in ASKA transfectants when applied before or after αTCR stimulation at 6 µM), and SykM442A S505A (56% inhibition of phagocytosis of Ab-opsonized SRBC by ASKA macrophages at 6 µM), while exhibiting little or no activity toward the wild-type or other off-target cellular enzymes. Functional analog-sensitive allele mutants are valuable tools in pharmacological validation of drug development and in elucidating enzyme signalings and functions both in cultures in vitro and in animals in vivo when specific inhibitors against naturally occurring target enzymes are unavailable and/or generations of knockout animals/cells lacking the target enzymes appear to be difficult, infeasible, or simply impossible.
A cell-permeable C3-enlarged PP1 (Cat. No. 567809) analog that is shown to act as a potent, ATP-competitive, and highly ASKA- (analog-sensitive kinase alleles) selective inhibitor against polo-like kinase 1C67V L130G (75% and 94% inhibition in cell-free kinase assays at 0.1 and 1 µM, respectively), ZAP-70C405V M414A/ZAP-70M414A (complete prevention and elimination, respectively, of αTCR-stimulated [Ca2+] elevation in ASKA transfectants when applied before or after αTCR stimulation at 6 µM), and SykM442A S505A (56% inhibition of phagocytosis of Ab-opsonized SRBC by ASKA macrophages at 6 µM), while exhibiting little or no activity toward the wild-type or other off-target cellular enzymes. Functional analog-sensitive allele mutants are valuable tools in pharmacological validation of drug development and in elucidating enzyme signalings and functions both in cultures in vitro and in animals in vivo when specific inhibitors against naturally occurring target enzymes are unavailable and/or generations of knockout animals/cells lacking the target enzymes appear to be difficult, infeasible, or simply impossible.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Miller, A.L., et al. 2009. J. Immunol.182, 988.
Levin, S.E., et al. 2008. J. Biol. Chem.283, 15419.
Burkard, M.E., et al. 2007. Proc. Natl. Acad. Sci. USA104, 4383.
Bishop, A.C., et al. 1999. J. Am. Chem. Soc.121, 627.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Shortt et al.
Methods in molecular biology (Clifton, N.J.), 2071, 171-186 (2019-11-24)
Apicomplexans are obligate parasites that replicate inside host cells, within a subcellular compartment called the parasitophorous vacuole. Egress is the process by which apicomplexan parasites like Toxoplasma gondii exit from host cells, rupturing the parasitophorous vacuole and host-cell plasma membranes
Dunja Celebic et al.
The EMBO journal, 43(7), 1325-1350 (2024-02-07)
Exit from mitosis is brought about by dramatic changes in the phosphoproteome landscape. A drop in Cyclin-dependent kinase (Cdk) activity, the master regulatory kinase, and activation of counteracting phosphatases such as Cdc14 in budding yeast, results in ordered substrate dephosphorylation, allowing
Jarmila Princová et al.
PLoS genetics, 19(1), e1010582-e1010582 (2023-01-11)
Oxidative stress is associated with cardiovascular and neurodegenerative diseases, diabetes, cancer, psychiatric disorders and aging. In order to counteract, eliminate and/or adapt to the sources of stress, cells possess elaborate stress-response mechanisms, which also operate at the level of regulating
Alex W Chan et al.
eLife, 12 (2023-11-07)
Apicomplexan parasites use Ca2+-regulated exocytosis to secrete essential virulence factors from specialized organelles called micronemes. Ca2+-dependent protein kinases (CDPKs) are required for microneme exocytosis; however, the molecular events that regulate trafficking and fusion of micronemes with the plasma membrane remain
Nadine Krämer et al.
Viruses, 14(10) (2022-10-28)
The human cytomegalovirus (HCMV) is a member of the beta-herpesvirus family and inflicts life-long latent infections in its hosts. HCMV has been shown to manipulate and dysregulate many cellular processes. One major interactor with the cellular host is the viral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service