Skip to Content
Merck

528119

PI 3-K Inhibitor XI, HWT

The PI 3-K Inhibitor XI, HWT, also referenced under CAS 58053-83-1, controls the biological activity of PI 3-K. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-K Inhibitor XI, HWT, Wortmannin, 17β-Hydroxy, LY 301497, mTOR Inhibitor VIII

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H26O8
CAS Number:
Molecular Weight:
430.45
UNSPSC Code:
12352200
Assay:
≥97% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 100 mg/mL (Use only fresh DMSO.)

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable Wortmannin (Cat. Nos. 681675 and 681676) 17β-Hydroxy analog (HWT) that acts as an irreversible PI 3-K inhibitor with an 8.4-fold higher potency than that of Wortmannin (IC50 = 0.5 and 4.2 nM, respectively) and inhibits mTOR only at much higher concentrations (IC50 = 0.193 µM). Both Wortmannin and HWT covalently modify a conserved lysine residue (Lys833 in porcine PI 3-Kγ) in the PI 3-K ATP binding pocket via their electrophilic furan ring at C-20 position. HWT is shown to effectively arrest the growth of LNCaP cells (IC50 = 1.464 µM) in vitro, however, its high in vivo toxicity renders it unsuitable for tumor killing studies in mice. Also reported to inhibit late phase PLD-mediated PA production upon fMLP (Cat. No. 05-22-2500) stimulation of human neutrophils.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zask, A., et al. 2008. J. Med. Chem.51, 1319.
Zhu, T., et al. 2006. J. Med. Chem.49, 1373.
Norman, B. H., et al. 1996. J. Med. Chem.39, 1106.
Gelas, P., et al. 1992. Biochem. J.287, 67.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library