Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C26H24ClN3O3 · xHCl
Molecular Weight:
461.94 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.28
Assay:
≥99% (HPLC)
Form:
powder
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Product 526529 is not currently sold in your country. Contact Technical Service
Quality Segment
assay
≥99% (HPLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated (hygroscopic), protect from light
color
orange
solubility
DMSO: 25 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
O=C(/C1=C/C2=CC=C(C3=CC=CC(C(N4CCCN(C)CC4)=O)=C3)O2)NC5=C1C=C(Cl)C=C5.[xHCl]
General description
An orally bioavailable oxindolo-furanyl compound that acts as a potent, reversible and ATP-competitive inhibitor of pan PIM-kinases (IC50 = 5, 25 and 16 nM for PIM-1, PIM-2 and PIM-3, respectively; Ki = 5 nM for PIM-1) with excellent selectivity over 107 kinases (IC80 >500 nM). Shown to arrest the proliferation of several human cancer cells (IC50 = 20 and 452 nM for MV-4-11 and PC3 cells) and block phosphorylation of Bad-Ser112, 4E-BP1-Ser65 and Thr37/46 with an IC50 of ~ 1.0 µM. Displays favorable pharmacokinetic profile and efficacy in mice, and suppresses tumor growth in MV-4-11 and PC3 xenograft murine model (50 mg/kg, p.o.). Further, potentiates doxorubicin (Cat. No. 324380 ) and paclitaxel (Cat. No. 580555 ) toxicity in PC3 cells.
Biochem/physiol Actions
Cell permeable: yes
Primary Target
PIM Kimases
PIM Kimases
Reversible: yes
Secondary Target
Flt3
Flt3
Target IC50: 5, 25 and 16 nM for PIM-1, PIM-2 and PIM-3
Packaging
Packaged under inert gas
Preparation Note
Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 6 months at -70°C.
Other Notes
Haddach, M., et al. 2012. ACS Med. Chem. Lett.3, 135.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.