Skip to Content
Merck

Skip To

513100

DL-threo-PDMP, Hydrochloride

PDMP closely resembles the natural sphingolipid substrate of brain glucosyltransferase and acts as a potent and competitive inhibitor of this enzyme.

Synonym(s):

DL-threo-PDMP, Hydrochloride, 1-Phenyl-2-decanoylamino-3-morpholino-1-propanol, HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mg

Estimated to ship onJune 08, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 6,790.00

About This Item

Empirical Formula (Hill Notation):
C23H38N2O3 · xHCl
CAS Number:
Molecular Weight:
390.56 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Storage condition:
OK to freeze

CZK 6,790.00


Estimated to ship onJune 08, 2026Details


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: soluble, ethanol: soluble, methanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

Cl.N2(CCOCC2)C[C@@H](NC(=O)CCCCCCCCC)[C@H](O)c1ccccc1

InChI

1S/C23H38N2O3.ClH/c1-2-3-4-5-6-7-11-14-22(26)24-21(19-25-15-17-28-18-16-25)23(27)20-12-9-8-10-13-20;/h8-10,12-13,21,23,27H,2-7,11,14-19H2,1H3,(H,24,26);1H/t21-,23-;/m1./s1

InChI key

HVJHJOYQTSEKPK-BLDCTAJRSA-N

General description

PDMP closely resembles the natural sphingolipid substrate of brain glucosyltransferase and acts as a potent and competitive inhibitor of this enzyme. Blocks the outgrowth of neurites and inhibits glycolipid synthesis in cultured NIH/3T3 cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
glycolipid synthesis in cultured NIH/3T3 cells
Product does not compete with ATP.
Reversible: no

Other Notes

Rani, C.S., et al. 1995. J. Biol. Chem. 270, 2859.
Rosenwald, A.G. 1992. Biochemistry31, 3581.
Inokuchi, J., and Radin, N.S. 1987. J. Lipid Res.28, 565.
Vunnam, R.R., and Radin, N.S. 1980. Chem. Phys. Lipids26, 265.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
310500567727203291
assay

≥98% (HPLC)

assay

≥98% (TLC)

assay

≥95% (elemental analysis)

assay

≥95% (HPLC)

form

solid

form

oil (Clear), waxy solid

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

DMSO: soluble, methanol: soluble

solubility

DMSO: 5 mg/mL, ethanol: soluble, methanol: soluble

solubility

methanol: 0.5 mg/mL

solubility

water: 10 mg/mL, DMSO: 20 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters