Skip to Content
Merck

5.09306

Late Endosome Trafficking Inhibitor, EGA

Synonym(s):

Late Endosome Trafficking Inhibitor, EGA, 4-Bromobenzaldehyde-N-(2,6-dimethylphenyl)semicarbazone, ( E)-2-(4-Bromobenzylidene)-N-(2,6-dimethylphenyl)hydrazinecarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H16BrN3O
CAS Number:
Molecular Weight:
346.22
MDL number:
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥93% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥93% (HPLC)

Quality Segment

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

General description

A cell-permeable semicabazone compound that prevents B. anthrax Lethal toxin/LT (EC50 = 1.7 µM in RAW264.7 cultures; ECmax = 6 µM in murine BMDM cultures), diphtheria toxin/DT, P. aeruginosa ExoA, and H. ducreyi cytolethal distending toxin (Hd-CDT) from exerting their cytotoxic activity, but not endocytic uptake, by blocking endosomal trafficking/maturation into acidified late endosomes, while exhibiting no cytoprotective efficacy against ricin & E. coli Ec-CDT that do not require acidified endosomes for intoxication. Likewise, EGA is also shown to inhibit low-pH-dependent viral infectivity by Armstrong strain of LCMV, A/WSN/33 strain of influenza virus, but not low-pH-independent infection of Nipah virus NiV or MLV. Although the exact molecular target remains unknown, EGA does not act as a pH neutralizing agent, nor does it inhibit endosomal recycling or trafficking to phagocytic compartments.
A cell-permeable semicarbazone compound that prevents B. anthrax Lethal toxin/LT (Protective Antigen/PA & LF/Lethal Factor complex; EC50 = 1.7 µM in RAW264.7 cultures; ECmax = 6 µM in murine BMDM cultures), diphtheria toxin (DT), LF-DT A chain fusion LFnDTA/PA complex, P. aeruginosa ExoA, and H. ducreyi cytolethal distending toxin (Hd-CDT) from exerting their cytotoxic activity, but not endocytic uptake, by blocking endosomal trafficking/maturation into acidified late endosomes, while exhibiting no cytoprotective efficacy against toxins that do not require acidified endosomes for intoxication, such as ricin & E. coli Ec-CDT. Likewise, EGA is also shown to inhibit low-pH-dependent viral infectivity by Armstrong strain of LCMV, A/WSN/33 strain of influenza virus, vesicular stomatitis virus VSV-pseudotyped lentivirus & VSV-pseudotyped MLV, but not low-pH-independent infection of Nipah virus NiV or MLV. Although the exact molecular target remains unknown, EGA does not act as a pH neutralizing agent, nor does it inhibit endosomal recycling or trafficking to phagocytic compartments.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
host-membrane trafficking
Reversible: yes

Packaging

Packaged under inert gas

Preparation Note

Use only fresh DMSO for reconstitution.

Other Notes

Gillespie, E.J., et al. 2013. Proc. Natl. Acad. Sci. USA110, E4904.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library