Skip to Content
Merck
All Photos(1)

Documents

5.08320

Sigma-Aldrich

Ezh2 Inhibitor III, SAH-EZH2

Synonym(s):

Ezh2 Inhibitor III, SAH-EZH2, H3K27 Methylation Inhibitor, SAH-EZH2, EED-EZH2 Protein-Protein Interaction Inhibitor, SAH-EZH2, Enhancer of Zested Homolog 2 Inhibitor III, HMTase Inhibitor XIV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C155H256N48O40 · 6CF3CO2H · XXH2O
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

potency

320 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 100 mg/mL

storage temp.

−20°C

General description

Ezh2 Inhibitor III, SAH-EZH2 is a stabilizeda-helical 27-mer peptide (FSSNRXKILXRTQILNQEWKQRRIQPV )with E54Q modification. It shows strong cellular uptake and nuclearlocalization and directly forms a complex during embryonic ectodermdevelopment. It blocks the protein-protein interaction of embryonic ectoderm development(EED) with enhancer of zeste homolog (EZH) 1 and 2. Selectively inhibits methyltransferase activity of the Polycombrepressive complex 2 (PRC2) and reduces H3K27 methylation in a dose-dependentmanner (~1 to 10 µM). This inhibitor induces growth arrest after six days oftreatment at the G0/G1 phase and monocyte-macrophage differentiation in MLL-AF9leukemia cells without inducing any apoptosis. The growth arrest is suggestedto be associated with an increase in p19ARF levels. It blocks the proliferationof M1 myeloid leukemia cells and C1498 without having any effect onthe non-tumorigenic HPC5 cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
EED (embryonic ectoderm development) - EZH2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-FSSNRXKILXRTQILNQEWKQRRIQPV (X=(S)-2-(4ʹ-pentenyl)Ala, stapled)

Other Notes

Kim, W., et al. 2013. Nat. Chem. Biol.9, 643.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly H Kim et al.
Nature medicine, 22(2), 128-134 (2016-02-06)
Recent genomic studies have resulted in an emerging understanding of the role of chromatin regulators in the development of cancer. EZH2, a histone methyl transferase subunit of a Polycomb repressor complex, is recurrently mutated in several forms of cancer and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service