Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C23H21FN6O
CAS Number:
Molecular Weight:
416.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Product Name
PERK Inhibitor II, GSK2656157,
assay
≥98% (HPLC)
Quality Segment
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
white
solubility
DMSO: 100 mg/mL
storage temp.
−20°C
SMILES string
Fc1c2c(ccc1c4c5c([n](c4)C)ncnc5N)N(CC2)C(=O)Cc3nc(ccc3)C
InChI key
PRWSIEBRGXYXAJ-UHFFFAOYSA-N
General description
A cell-permeable pyrrolopyrimidinamine compound that acts as a potent, ATP-competitive EIF2AK3/PERK inhibitor (IC50 = 0.9 nM; [ATP] = 10 µM), displaying ≥511-fold selectivity over HRI/EIF2AK1, BRK, PKR/EIF2AK2, and MEKK2 (IC50 = 460, 822, 905, and 954 nM, respectively) and much reduced or little activity toward more than 300 other kinases. While GSK2656157 potently inhibits ER stress-induced enhancement of PERK and eIF2α phosphorylation as well as ATF4 & CHOP upregulation in human PxBC3 and murine LL/2 cultures in vitro (IC50 ≤30 nM; 1 h drug preincubation prior to 6 h ER-stress induction by 1 µM Thapsigargin, only basal PERK phosphorylation, but not basal eIF2α phosphorylation or ATF4 & CHOP expression, is seen inhibited in pancreas (by >95% up to 8 h post single 50 mg/kg oral dose) and tumor tissues upon GSK2656157 administration in mice in vivo, resulting in the observed antitumor efficacy (54-110% tumor growth inhibition; 150 mg/kg/12 h via p.o.) and reversible pancreas damage (44% and 95% of control pancreas mass, respectively, 1 d and 15 d post 14 d 150 mg/kg/12 h oral dosing). Comparing to its structural analog GSK2606414 (Cat. No. 516535), GSK2656157 exhibits much reduced potency toward Aurora B, MLK2/MAP3K10, MLCK2/MYLK2, c-MER (IC50 = 1.259, 2.796, 3.039, and 3.431 µM, respectively), c-Kit, and DDR2 (69.84% and 25.41% inhibition, respectively, at 10 µM)
Biochem/physiol Actions
Cell permeable: yes
Primary Target
PERK
PERK
Reversible: yes
Packaging
Packaged under inert gas
Preparation Note
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.
Other Notes
Atkins, C., et al. 2013. Cancer Res.73, 1993.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Regulatory Review (Z)
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.