Skip to Content
Merck
All Photos(1)

Key Documents

5.00527

Sigma-Aldrich

APETx2, Anthopleura elegantissima, Recombinant, E. coli

recombinant, expressed in E. coli

Synonym(s):

APETx2, Anthopleura elegantissima, Recombinant, E. coli, Acid-Sensing Ion Channel 3 Blocker, ASIC3 Channel Blocker

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C196H280N54O61S6
CAS Number:
Molecular Weight:
4561.04
UNSPSC Code:
12352200
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder

potency

63 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

aqueous buffer: soluble

storage temp.

−20°C

General description

Originally isolated from sea anemone Anthopleura elegantissima, the 42 amino-acid toxin APETx2 is shown to block acidification-induced activation of homotrimeric acid-sensing ion channel composed of rat or human ASIC3 (IC50 against pH 7.4 to 6 ramping-induced transient current = 63 nM and 175 nM by whole-cell patch-clamp using COS cells expressing respective channel), but not rat ASIC1a, ASIC1b, or ASIC2a, in a reversible manner, presumably via its N-terminus interaction with type I β turn that connects β-strands I&II in the ASIC3 extracellular β-ball region. In addition to acidification-induced classical transient current, APETx2 is also shown to inhibit the Na+-dependent, alkalization-induced nonconventional channel activity seen in human, but not rat, ASIC3 (complete inhibition by 1 µM APETx3; pH 8.0). APETx2 in vivo efficacy is demonstrated in various rat pain induction models, including acid- (via calf i.m. injection of 100 µL pH 4.0 saline on day 1&5) induced mechanical hypersensitivity (pain withdrawal threshold = 76% and 24% of control rats with or without 100 µL 2.2 µM APETx2 i.m. injection prior to the second pH 4.0 saline injection). Although structurally related to known K+ channel modulators APETx1 and BDS-1/II, APETx2 is reported to display much reduced potency against Kv3.4 (38% inhibition by 3 µM APETx2) and little or no effect toward hERG, Kv2.2, Kv3.1, Kv4.2, or Kv4.3 channel activity.

Biochem/physiol Actions

Primary Target
ASIC3
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Gly-Thr-Ala-Cys⁴-Ser-Cys⁶-Gly-Asn-Ser-Lys-Gly-Ile-Tyr-Trp-Phe-Tyr-Arg-Pro-Ser-Cys²⁰-Pro-Thr-Asp-Arg-Gly-Tyr-Thr-Gly-Ser-Cys³⁰-Arg-Tyr-Phe-Leu-Gly-Thr-Cys³⁷-Cys³⁸-Thr-Pro-Ala-Asp-OH (disulfide bonds: 4 → 37, 6 → 30, 20 → 38)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Delaunay, A., et al. 2012. Proc. Natl. Acad. Sci.109, 13124.
Sherwood, T.W., et al. 2012. Am. J. Physiol. Cell Physiol.303, C699.
Karczewski, J., et al. 2010. Br J Pharmacol.161, 950.
Chagot, B., et al. 2005. Protein Sci.14, 2003.

Diochot, S., et al. 2004. EMBO J.23, 1516.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service