Skip to Content
Merck
All Photos(2)

Key Documents

489478

Sigma-Aldrich

LSD1 Inhibitor III, CBB1007

The LSD1 Inhibitor III, CBB1007 controls the biological activity of LSD1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

LSD1 Inhibitor III, CBB1007, LSD Inhibitor III, BHC110 Inhibitor III, Methyl-3-(4-(4-carbamimidoylbenzoyl)piperazine-1-carbonyl)-5-((4-carbamimidoylpiperazin-1-yl)methyl)benzoate, 3TFA, KDM1 Inhibitor III, Histone Lysine Demethylase Inhibitor V, LSD Inhibitor III, Methyl-3-(4-(4-carbamimidoylbenzoyl)piperazine-1-carbonyl)-5-((4-carbamimidoylpiperazin-1-yl)methyl)benzoate, 3TFA, KDM1 Inhibitor III, Histone Lysine Demethylase Inhibitor V, BHC110 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H34N8O4 · 3C2HF3O2
CAS Number:
Molecular Weight:
876.68
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

NC(N1CCN(CC2=CC(C(N3CCN(C(C4=CC=C(C(N)=N)C=C4)=O)CC3)=O)=CC(C(OC)=O)=C2)CC1)=N

General description

A cell-permeable amidino-guanidinium compound that acts as a potent, reversible and substrate competitive LSD1 selective inhibitor (IC50 = 5.27 µM for hLSD1) and efficiently blocks LSD1-mediated demethylation of H3K4Me2 and H3K4Me (IC50 ≤ 5 µM) with no effect on H3K4Me3 and H3K9Me2, and LSD2 and JARID1A activities. Increases H3K4Me2 and H3K4Me contents (IC50 ≤ 5 µM), and causes activation of epigenetically suppressed CHRM4/M4-ArchR and SCN3A genes in F9 cells (IC50 ≤ 3.74 µM). Shown to preferentially arrest the growth of pluripotent tumors with minimal effect on non-pluripotent cancer or normal somatic cells (IC50 >100 µM).

Note: the molecular weight is lot-specific and can be found on the vial label.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wang, J., et al. 2011. Cancer Res.71, 7238.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service