Skip to Content
Merck

Skip To

487952

NOC-7

A nitric oxide (NO) donor similar to NOC-5 but with a much shorter half-life of NO release.

Synonym(s):

NOC-7, 3-(2-Hydroxy-1-methyl-2-nitrosohydrazino)-N-methyl-1-propanamine

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C5H14N4O2
CAS Number:
Molecular Weight:
162.19
UNSPSC Code:
12352200
Assay:
≥90% (HPLC)
Form:
solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light


Quality Segment

assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

color

white

solubility

0.1 M NaOH: soluble, water: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C5H14N4O2/c1-6-4-3-5-8(2)9(11)7-10/h6,11H,3-5H2,1-2H3

InChI key

AXAGAUVQMYPYHM-UHFFFAOYSA-N

General description

A nitric oxide (NO) donor similar to NOC-5 (Cat. No. 487950) but with a much shorter half-life of NO release. Half-life (t½) in PBS, pH 7.4 is 10 minutes at 22°C. Useful as a short-acting nitrovasodilator. Relatively stable in alkaline solution (pH ≥10.0).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A nitric oxide (NO) donor
Product does not compete with ATP.
Reversible: no

Preparation Note

Following reconstitution in alkaline solution, aliquot and freeze (-20°C). Unstable in aqueous solutions; reconstitute in H₂O jut prior to use. Alkaline stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, P., et al. 1996. Circulation94, 2235.
Hrabie, J.A., et al. 1993. J. Org. Chem.58, 1472.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters