Skip to Content
Merck

Skip To

487052

p-Nitrophenyl-N-acetyl-β-D-glucosaminide

≥ 99% (TLC), solid, N-acetyl-β-glucosaminidase substrate, Calbiochem®

Synonym(s):

p-Nitrophenyl-N-acetyl-β-D-glucosaminide, p-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
CZK 5,150.00

About This Item

Empirical Formula (Hill Notation):
C14H18N2O8
CAS Number:
Molecular Weight:
342.30
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Form:
solid
Storage condition:
OK to freeze

CZK 5,150.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

p-Nitrophenyl-N-acetyl-β-D-glucosaminide, Useful substrate for rapid colorimetric assay of N-acetyl-β-glucosaminidase activity in human urine.

Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

0.1 M citrate buffer, pH 4.2: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1ccc(cc1)O[C@@H]2O[C@@H]([C@H]([C@@H]([C@H]2NC(=O)C)O)O)CO

InChI

1S/C14H18N2O8/c1-7(18)15-11-13(20)12(19)10(6-17)24-14(11)23-9-4-2-8(3-5-9)16(21)22/h2-5,10-14,17,19-20H,6H2,1H3,(H,15,18)/t10-,11-,12-,13-,14-/m1/s1

InChI key

OMRLTNCLYHKQCK-DHGKCCLASA-N

General description

Useful substrate for rapid colorimetric assay of N-acetyl-β-glucosaminidase activity in human urine.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Heating (37°C) may be required to completely solubilize.

Other Notes

Jung, K., and Becker, S. 1991. Biomed. Biochim. Acta50, 861.
Maruhn, D. 1976. Clin. Chim. Acta73, 453.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
487506487507487500
form

solid

form

solid

form

solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

10-30°C

storage temp.

2-8°C

solubility

0.1 M citrate buffer, pH 4.2: soluble

solubility

water: 10 mg/mL

solubility

water: soluble

solubility

water: 10 mg/mL

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze

storage condition

OK to freeze, protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters