Skip to Content
Merck
All Photos(1)

Key Documents

48-624MAG

Millipore

MILLIPLEX® 2-Plex Phospho/Total p38 - Cell Signaling Multiplex Assay

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
CZK 26,300.00

CZK 26,300.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
96 EA
CZK 26,300.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

CZK 26,300.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

species reactivity

mouse, human, rat

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Protein phosphorylation represents a key mechanism used in regulating cellular functions in all eukaryotic cells. Aberrant phosphorylation has been implicated in the onset and development of many diseases including metabolic disorders, inflammatory disease, cancer, etc. Changes in protein phosphorylation can be attributed to changes in both phosphorylation events, as well as changes due to total protein levels. To distinguish the changes in phosphorylation from changes in protein expression it is important to normalize the signal from phosphorylation over the signal from total protein. For this need, the MILLIPLEX® MAP 2-plex Phospho/Total p38 kit has been developed for the simultaneous detection of phosphorylated p38 (Thr180/Tyr182) and total p38 in a single well using the Luminex system. According to the original vendor′s antibody information sheets, phospho-p38 antibody pair detects p38α, p38β and p38γ but not p38-δ, while the total p38 antibody pair detects only p38α. The detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures for the analysis of cell lysate samples. Each kit has sufficient reagents for one 96 well plate assay.The MILLIPLEX® 2-Plex Phospho/Total p38 magnetic bead kit has been developed for the simultaneous detection of phosphorylated p38 (Thr180/Thr182) and total p38 in a single well using the Luminex® system. This detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures for the analysis of cell lysate samples.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation or total pathway proteins in tissue and cell lysate samples.An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted, filtered cell lysate per well.This kit must be run using the provided Assay Buffer.The suggested working range of protein concentration for the assay is 1 to 25 μg of total protein/well (25 μL/well at 40 to 1,000 μg/mL)Analytes available:p38 (Total);p38 (Thr180/Tyr182)

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Uncover how cells communicate with MILLIPLEX® cell signaling multiplex assays. Multiplexing with cell signaling phosphoprotein assays based on Luminex® xMAP® technology helps researchers measure phosphoproteins and total proteins within the same or different pathways from a single sample.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service