Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
Estimated to ship onJune 01, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 4,250.00 |
About This Item
Quality Segment
assay
≥98% (TLC)
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
white
solubility
DMSO: 50 mg/mL, methanol: 50 mg/mL
storage temp.
2-8°C
SMILES string
[Mg+2].N([C@@H](CCC(=O)[O-])C(=O)[O-])C(=O)[C@@H](NC(=O)C)CC(=O)[O-].[H+]
InChI
1S/C11H16N2O8.Mg/c1-5(14)12-7(4-9(17)18)10(19)13-6(11(20)21)2-3-8(15)16;/h6-7H,2-4H2,1H3,(H,12,14)(H,13,19)(H,15,16)(H,17,18)(H,20,21);/q;+2/p-2/t6-,7-;/m0./s1
InChI key
BQAXDBLSAIPBND-LEUCUCNGSA-L
General description
Preparation Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (TLC) | assay ≥99% (HPLC) | assay ≥99% (HPLC) | assay - |
| form powder | form - | form powder | form - |
| manufacturer/tradename Calbiochem® | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename Upstate® |
| Quality Level 100 | Quality Level 300 | Quality Level 300 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. - | storage temp. - | storage temp. - |
| solubility DMSO: 50 mg/mL, methanol: 50 mg/mL | solubility - | solubility - | solubility - |
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




