Skip to Content
Merck

475916

MOPS/EDTA Buffer, 10X Liquid Concentrate, Molecular Biology Grade - Calbiochem

Prepared with DEPC for RNA applications. 10X concentrate contains 200 mM MOPS, 50 mM sodium acetate, 10 mM EDTA, pH 7.0 at 25°C.

Synonym(s):

MOPS/EDTA Buffer, 10X Liquid Concentrate, Molecular Biology Grade - Calbiochem

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H15NO4S
Molecular Weight:
209.26
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

solubility

water: soluble

foreign activity

DNases, RNases, none detected

shipped in

ambient

storage temp.

10-30°C

General description

Prepared with DEPC for RNA applications. 10X concentrate contains 200 mM MOPS, 50 mM sodium acetate, 10 mM EDTA, pH 7.0 at 25°C.
Prepared with diethylpyrocarbonate (DEPC) for RNA applications. 10X concentrate contains 200 mM MOPS, 50 mM sodium acetate, 10 mM EDTA, pH 7.0 at 25°C. The most commonly used buffer for RNA electrophoresis in denaturing (formaldehyde) agarose gels.

Other Notes

Ogden, R.C., and Adams, D.A. 1987. Methods Enzymol.152, 61.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library